Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327883.1 | NP_001314812.1 | 4 - 5 | 460 | 69 | |
GenBank mRNA | BC079485.1 | - | 4 - 5 | 431 | 69 | |
JN242071.1 | - | 4 - 5 | 359 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |