Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327883.1 | NP_001314812.1 | 5 - 6 | 614 | 107 | |
GenBank mRNA | BC079485.1 | - | 5 - 6 | 585 | 107 | |
JN242071.1 | - | 5 - 6 | 513 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |