Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327883.1 | NP_001314812.1 | 8 - 9 | 1038 | 78 | |
GenBank mRNA | BC079485.1 | - | 8 - 9 | 1009 | 78 | |
JN242071.1 | - | 8 - 9 | 937 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |