Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003991.1 | NP_001003991.1 | 5 - 6 | 359 | 82 | |
GenBank mRNA | BC080262.1 | - | 5 - 6 | 359 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |