Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005586.2 | NP_001005586.1 | 4 - 5 | 739 | 63 | |
XM_005165122.4 | XP_005165179.1 | 5 - 6 | 759 | 63 | ||
XM_005165123.4 | XP_005165180.1 | 5 - 6 | 756 | 63 | ||
XM_005165124.4 | XP_005165181.1 | 4 - 5 | 648 | 63 | ||
GenBank mRNA | AY070260.1 | - | 4 - 5 | 580 | 63 | |
AY070262.1 | - | 5 - 6 | 682 | 63 | ||
AY070263.1 | - | 4 - 5 | 739 | 63 | ||
BC081573.1 | - | 4 - 5 | 584 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |