Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008608.1 | NP_001008608.1 | 4 - 5 | 599 | 83 | |
GenBank mRNA | BC086819.1 | - | 4 - 5 | 599 | 83 | |
BC164266.1 | - | 4 - 5 | 550 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |