Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017636.1 | NP_001017636.1 | 5 - 6 | 700 | 132 | |
GenBank mRNA | BC092815.1 | - | 5 - 6 | 700 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |