Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fsip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017876.1 | NP_001017876.1 | 1390 | 105 | ||
GenBank mRNA | BC093429.1 | - | 1390 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio |