Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep55l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020762.1 | NP_001018598.1 | 4 - 5 | 590 | 54 | |
XM_005156020.4 | XP_005156077.1 | 4 - 5 | 605 | 54 | ||
GenBank mRNA | BC095686.1 | - | 4 - 5 | 590 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |