Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025177.1 | NP_001020348.1 | 5 - 6 | 622 | 124 | |
XM_005163623.4 | XP_005163680.1 | 5 - 6 | 663 | 124 | ||
GenBank mRNA | BC097131.1 | - | 5 - 6 | 622 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |