Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030116.1 | NP_001025287.1 | 5 - 6 | 798 | 92 | |
XM_005160433.1 | XP_005160490.1 | 5 - 6 | 854 | 92 | ||
XM_005160434.1 | XP_005160491.1 | 5 - 6 | 854 | 92 | ||
GenBank mRNA | BC124746.1 | - | 5 - 6 | 801 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |