Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glt8d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039931.1 | NP_001035020.1 | 5 - 6 | 609 | 66 | |
GenBank mRNA | BC114297.1 | - | 5 - 6 | 609 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |