Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: antxr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044709.2 | NP_001038174.2 | 11 - 12 | 1260 | 140 | |
GenBank mRNA | BC139636.1 | - | 11 - 12 | 1260 | 140 | |
DQ415957.1 | - | 11 - 12 | 915 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |