Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: antxr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044709.2 | NP_001038174.2 | 9 - 10 | 1107 | 99 | |
GenBank mRNA | BC139636.1 | - | 9 - 10 | 1107 | 99 | |
DQ415957.1 | - | 9 - 10 | 762 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |