Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dvl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044917.1 | NP_001038382.1 | 14 - 15 | 1636 | 75 | |
XM_005169297.4 | XP_005169354.1 | 13 - 14 | 2132 | 75 | ||
XM_005169298.4 | XP_005169355.1 | 14 - 15 | 2054 | 75 | ||
XM_005169299.4 | XP_005169356.1 | 11 - 12 | 1950 | 75 | ||
GenBank mRNA | BC171461.1 | - | 14 - 15 | 1745 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |