Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044937.2 | NP_001038402.1 | 19 - 20 | 2954 | 72 | |
XM_005170050.4 | XP_005170107.1 | 20 - 21 | 3634 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |