Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p4ha1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044998.1 | NP_001038463.1 | 10 - 11 | 1210 | 65 | |
XM_009306501.3 | XP_009304776.1 | 10 - 11 | 1413 | 65 | ||
XM_009306502.3 | XP_009304777.1 | 10 - 11 | 1413 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |