Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snapin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045148.1 | NP_001038613.1 | 1 - 2 | 117 | 82 | |
GenBank mRNA | BC139575.1 | - | 1 - 2 | 145 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |