Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045214.1 | NP_001038679.1 | 22 - 23 | 2726 | 97 | |
XM_005170062.4 | XP_005170119.1 | 22 - 23 | 3137 | 97 | ||
GenBank mRNA | BC163250.1 | - | 21 - 22 | 2684 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |