Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045419.2 | NP_001038884.2 | 6 - 7 | 805 | 65 | |
NM_001139438.1 | NP_001132910.1 | 5 - 6 | 965 | 65 | ||
GenBank mRNA | BC122098.1 | - | 6 - 7 | 800 | 65 | |
CF348078.1 | - | 2 - 3 | 187 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |