Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075103.1 | NP_001068571.1 | 3 - 4 | 1032 | 102 | |
GenBank mRNA | BC096901.1 | - | 2 - 3 | 313 | 102 | |
BC124299.1 | - | 3 - 4 | 1032 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |