Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075103.1 | NP_001068571.1 | 6 - 7 | 1329 | 77 | |
GenBank mRNA | BC096901.1 | - | 5 - 6 | 610 | 77 | |
BC124299.1 | - | 6 - 7 | 1329 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |