Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hs3st1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081439.1 | NP_001074908.1 | 646 | 65 | ||
GenBank mRNA | BC163410.1 | - | 736 | 65 | ||
BC163431.1 | - | 736 | 65 | |||
DQ812992.1 | - | 646 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |