Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vcam1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083835.1 | NP_001077304.1 | 6 - 7 | 1478 | 77 | |
XM_017353155.2 | XP_017208644.1 | 8 - 9 | 1989 | 77 | ||
XM_017353156.2 | XP_017208645.1 | 9 - 10 | 1811 | 77 | ||
GenBank mRNA | BC133177.1 | - | 6 - 7 | 1484 | 77 | |
CU458903.1 | - | 6 - 7 | 1478 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |