Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nptx2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109768.1 | NP_001103238.1 | 3 - 4 | 1399 | 67 | |
XM_005164291.4 | XP_005164348.1 | 3 - 4 | 2121 | 67 | ||
XM_005164292.4 | XP_005164349.1 | 3 - 4 | 2117 | 67 | ||
GenBank mRNA | BC076490.1 | - | 3 - 4 | 951 | 67 | |
BC115228.1 | - | 3 - 4 | 1399 | 67 | ||
FJ232033.1 | - | 3 - 4 | 896 | 67 | ||
KM036426.1 | - | 4 - 5 | 1797 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |