Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131159.2 | NP_571234.2 | 7 - 8 | 1132 | 119 | |
XM_017358977.2 | XP_017214466.1 | 7 - 8 | 1449 | 119 | ||
XM_017358978.2 | XP_017214467.1 | 7 - 8 | 1244 | 119 | ||
XM_017358979.2 | XP_017214468.1 | 7 - 8 | 1427 | 119 | ||
XM_017358980.2 | XP_017214469.1 | 7 - 8 | 1446 | 119 | ||
GenBank mRNA | U84615.1 | - | 7 - 8 | 1138 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |