Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131375.2 | NP_571450.1 | 8 - 9 | 663 | 65 | |
GenBank mRNA | AF195052.1 | - | 8 - 9 | 664 | 65 | |
BC075864.1 | - | 8 - 9 | 677 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |