Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131681.2 | NP_571756.1 | 2 - 3 | 411 | 83 | |
GenBank mRNA | AF329850.1 | - | 2 - 3 | 380 | 83 | |
BC097086.1 | - | 2 - 3 | 411 | 83 | ||
EH550887.1 | - | 2 - 3 | 448 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |