Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131681.2 | NP_571756.1 | 3 - 4 | 516 | 116 | |
NR_131978.1 | 2 - 3 | 1735 | 116 | |||
GenBank mRNA | AF329850.1 | - | 3 - 4 | 485 | 116 | |
BC081572.1 | - | 2 - 3 | 1673 | 116 | ||
EH476941.1 | - | 3 - 4 | 691 | 116 | ||
EH550887.1 | - | 3 - 4 | 553 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |