Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131722.1 | NP_571797.1 | 11 - 12 | 1254 | 68 | |
XM_005167061.4 | XP_005167118.1 | 11 - 12 | 1385 | 68 | ||
GenBank mRNA | AB030897.1 | - | 11 - 12 | 1254 | 68 | |
BC044128.1 | - | 11 - 12 | 1089 | 68 | ||
BC059417.1 | - | 11 - 12 | 1259 | 68 | ||
BC164402.1 | - | 11 - 12 | 1048 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |