Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152973.2 | NP_694505.2 | 1 - 2 | 571 | 57 | |
XM_021469720.1 | XP_021325395.1 | 1 - 2 | 684 | 57 | ||
GenBank mRNA | AF397015.1 | - | 1 - 2 | 562 | 57 | |
BC044348.1 | - | 1 - 2 | 461 | 57 | ||
BC061708.1 | - | 1 - 2 | 571 | 57 | ||
BC065358.1 | - | 1 - 2 | 565 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |