Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173221.2 | NP_775328.2 | 3 - 4 | 392 | 63 | |
XM_005163981.4 | XP_005164038.1 | 3 - 4 | 436 | 63 | ||
GenBank mRNA | AF254955.1 | - | 2 - 3 | 286 | 63 | |
BC049338.1 | - | 3 - 4 | 392 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |