Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197933.2 | NP_922915.2 | 12 - 13 | 1286 | 61 | |
GenBank mRNA | AF387900.1 | - | 12 - 13 | 1333 | 61 | |
BC044507.1 | - | 12 - 13 | 1286 | 61 | ||
BC165811.1 | - | 12 - 13 | 1294 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |