Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197933.2 | NP_922915.2 | 4 - 5 | 373 | 118 | |
GenBank mRNA | AF387900.1 | - | 4 - 5 | 420 | 118 | |
BC044507.1 | - | 4 - 5 | 373 | 118 | ||
BC165811.1 | - | 4 - 5 | 381 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |