Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: figla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198919.2 | NP_944601.2 | 1 - 2 | 376 | 128 | |
GenBank mRNA | AY302567.1 | - | 1 - 2 | 230 | 128 | |
BC092676.1 | - | 1 - 2 | 376 | 128 | ||
BC164317.1 | - | 1 - 2 | 264 | 128 | ||
EU723768.1 | - | 1 - 2 | 322 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |