Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199578.1 | NP_955872.1 | 8 - 9 | 943 | 78 | |
GenBank mRNA | BC059660.1 | - | 8 - 9 | 943 | 78 | |
BC071334.1 | - | 8 - 9 | 939 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |