Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpd52l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199582.2 | NP_955876.2 | 5 - 6 | 670 | 98 | |
NM_200885.2 | NP_957179.1 | 4 - 5 | 622 | 98 | ||
XM_005162032.2 | XP_005162089.1 | 7 - 8 | 170 | |||
XM_005162033.3 | XP_005162090.1 | 7 - 8 | 167 | |||
XM_005162035.2 | XP_005162092.1 | 6 - 7 | 170 | |||
XM_005162036.3 | XP_005162093.1 | 6 - 7 | 167 | |||
XM_005162037.2 | XP_005162094.1 | 5 - 6 | 140 | |||
XM_021469740.1 | XP_021325415.1 | 8 - 9 | 170 | |||
XM_021469741.1 | XP_021325416.1 | 8 - 9 | 167 | |||
XM_021469742.1 | XP_021325417.1 | 7 - 8 | 140 | |||
XM_021469744.1 | XP_021325419.1 | 6 - 7 | 745 | 98 | ||
GenBank mRNA | BC045297.1 | - | 5 - 6 | 650 | 98 | |
BC063969.1 | - | 4 - 5 | 588 | 98 | ||
EH444392.1 | - | 5 - 6 | 679 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |