Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: add3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199663.1 | NP_955957.1 | 9 - 10 | 1036 | 109 | |
XM_005161804.4 | XP_005161861.1 | 6 - 7 | 1042 | 109 | ||
XM_005161805.4 | XP_005161862.1 | 6 - 7 | 1042 | 109 | ||
XM_005161807.4 | XP_005161864.1 | 6 - 7 | 1041 | 109 | ||
XM_017353146.2 | XP_017208635.1 | 6 - 7 | 1043 | 109 | ||
XM_017353147.2 | XP_017208636.1 | 7 - 8 | 913 | 109 | ||
XM_017353148.2 | XP_017208637.1 | 6 - 7 | 1261 | 109 | ||
XM_017353149.2 | XP_017208638.1 | 6 - 7 | 1043 | 109 | ||
XM_017353150.2 | XP_017208639.1 | 6 - 7 | 1042 | 109 | ||
XM_021469329.1 | XP_021325004.1 | 8 - 9 | 1010 | 109 | ||
XM_021469330.1 | XP_021325005.1 | 6 - 7 | 771 | 109 | ||
XM_021469331.1 | XP_021325006.1 | 8 - 9 | 1009 | 109 | ||
GenBank mRNA | BC045384.1 | - | 6 - 7 | 940 | 109 | |
BC050486.1 | - | 9 - 10 | 1036 | 109 | ||
BC165643.1 | - | 5 - 6 | 757 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |