Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199723.1 | NP_956017.1 | 3 - 4 | 369 | 79 | |
GenBank mRNA | BC053252.1 | - | 3 - 4 | 369 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |