Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp1aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328560.1 | NP_001315489.1 | 1 - 2 | 173 | 94 | |
NM_001328561.1 | NP_001315490.1 | 1 - 2 | 173 | 94 | ||
NM_199945.2 | NP_956239.1 | 1 - 2 | 173 | 94 | ||
GenBank mRNA | BC059682.1 | - | 1 - 2 | 71 | 94 | |
EH446162.1 | - | 1 - 2 | 134 | 94 | ||
EH538724.1 | - | 1 - 2 | 212 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |