Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: washc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200183.1 | NP_956477.1 | 17 - 18 | 2759 | 113 | |
GenBank mRNA | BC045490.1 | - | 17 - 18 | 2759 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |