Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgap4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200444.1 | NP_956738.1 | 2 - 3 | 117 | 76 | |
XM_005162155.4 | XP_005162212.1 | 2 - 3 | 217 | 76 | ||
GenBank mRNA | BC054931.1 | - | 2 - 3 | 117 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |