Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200458.2 | NP_956752.2 | 3 - 4 | 453 | 110 | |
GenBank mRNA | AY391422.1 | - | 3 - 4 | 453 | 110 | |
BC045488.1 | - | 3 - 4 | 484 | 110 | ||
BC063965.1 | - | 3 - 4 | 464 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |