Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200670.1 | NP_956964.1 | 13 - 14 | 2390 | 85 | |
XM_005158332.4 | XP_005158389.1 | 12 - 13 | 2775 | 85 | ||
XM_005158333.4 | XP_005158390.1 | 12 - 13 | 3003 | 85 | ||
XM_005158334.4 | XP_005158391.1 | 14 - 15 | 2769 | 85 | ||
XM_005158335.4 | XP_005158392.1 | 11 - 12 | 2641 | 85 | ||
XM_009292432.3 | XP_009290707.1 | 14 - 15 | 3269 | 85 | ||
XM_009292434.3 | XP_009290709.1 | 13 - 14 | 2637 | 85 | ||
XM_021466650.1 | XP_021322325.1 | 14 - 15 | 2717 | 85 | ||
GenBank mRNA | BC058052.1 | - | 13 - 14 | 2390 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |