Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328361.1 | NP_001315290.1 | 4 - 5 | 661 | 79 | |
NM_201072.2 | NP_957366.1 | 3 - 4 | 616 | 79 | ||
XM_021477447.1 | XP_021333122.1 | 5 - 6 | 3327 | 79 | ||
GenBank mRNA | AY555037.1 | - | 3 - 4 | 604 | 79 | |
AY555038.1 | - | 4 - 5 | 649 | 79 | ||
BC049315.1 | - | 3 - 4 | 604 | 79 | ||
BC062828.1 | - | 3 - 4 | 616 | 79 | ||
EH454794.1 | - | 3 - 4 | 633 | 79 | ||
EH455188.1 | - | 4 - 5 | 658 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |