Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213048.3 | NP_998213.2 | 2 - 3 | 718 | 111 | |
GenBank mRNA | BC050494.1 | - | 2 - 3 | 705 | 111 | |
BC075760.1 | - | 2 - 3 | 719 | 111 | ||
BC165299.1 | - | 1 - 2 | 642 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |