Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213104.1 | NP_998269.1 | 1 - 2 | 427 | 65 | |
GenBank mRNA | BC053415.1 | - | 1 - 2 | 427 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |