Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214696.1 | NP_999861.1 | 4 - 5 | 497 | 117 | |
XM_009294525.1 | XP_009292800.1 | 4 - 5 | 561 | 117 | ||
XM_009294526.1 | XP_009292801.1 | 4 - 5 | 561 | 117 | ||
XM_009294527.1 | XP_009292802.1 | 3 - 4 | 390 | 117 | ||
GenBank mRNA | BC056750.1 | - | 4 - 5 | 497 | 117 | |
BC068373.1 | - | 4 - 5 | 494 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |