Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002485.2 | NP_001002485.1 | 2 - 3 | 304 | 82 | |
GenBank mRNA | BC076324.1 | - | 2 - 3 | 305 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |