Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002719.1 | NP_001002719.1 | 8 - 9 | 838 | 90 | |
XM_017356062.2 | XP_017211551.1 | 8 - 9 | 800 | 90 | ||
GenBank mRNA | BC075765.1 | - | 8 - 9 | 838 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |